Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thumpd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145585.2 | NP_663560.1 | 2 | 379 | ||
GenBank mRNA | AK032078.1 | - | 2 | 355 | ||
AK084107.1 | - | 2 | 349 | |||
AK152912.1 | - | 2 | 352 | |||
AK183558.1 | - | 1 | 99 | |||
BC004776.1 | - | 2 | 346 | 3257051 |