Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abca16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278943.1 | NP_001265872.1 | 9 | 1358 | ||
NM_001278944.1 | NP_001265873.1 | 12 | 1827 | |||
GenBank mRNA | AK144723.1 | - | 9 | 1359 | ||
AK161253.1 | - | 1 | 110 | |||
AY243472.1 | - | 12 | 1826 |