Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cog7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033318.3 | NP_001028490.2 | 4 | 620 | ||
GenBank mRNA | AK030709.1 | - | 4 | 587 | ||
AK145163.1 | - | 4 | 618 | |||
AK154693.1 | - | 4 | 622 | |||
AK163143.1 | - | 640 | ||||
AK172200.1 | - | 4 | 622 |