Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atxn2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183020.1 | NP_898841.1 | 3 | 455 | ||
GenBank mRNA | AK084800.1 | - | 3 | 526 | ||
AK155062.1 | - | 3 | 520 | |||
AK158626.1 | - | 3 | 142 | |||
AK168745.1 | - | 3 | 172 | |||
BC054483.1 | - | 3 | 455 | 6391591 |