Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PODXL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018111.2 | NP_001018121.1 | 6, 7 | 1501 | ||
NM_005397.3 | NP_005388.2 | 5, 6 | 1405 | |||
GenBank mRNA | AK055816.1 | - | 6, 7 | 1381 | ||
AK223573.1 | - | 5, 6 | 1397 | |||
BC042466.1 | - | 4, 5 | 1693 | |||
BC093730.1 | - | 5, 6 | 1167 | 7939575 | ||
BC112035.1 | - | 5, 6 | 1179 | 8327503 | ||
BC143318.1 | - | 6, 7 | 1269 | 9051823 | ||
CN290641.1 | - | 5, 6 | 593 | |||
U97519.1 | - | 5, 6 | 1403 |