Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arglu1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176849.3 | NP_789819.2 | 4 | 932 | ||
GenBank mRNA | AK169518.1 | - | 2 | 663 | ||
BC029219.1 | - | 2 | 2615 | |||
BC046971.1 | - | 4 | 3143 | 6468505 | ||
BU514203.1 | - | 5 | 579 |