Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmco3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172282.2 | NP_758486.1 | 5, 6 | 1392 | ||
GenBank mRNA | AK033499.1 | - | 5, 6 | 1153 | ||
AK046065.1 | - | 5, 6 | 1182 | |||
AK049974.1 | - | 5, 6 | 1151 | |||
AK088866.1 | - | 5, 6 | 1176 | |||
AK148252.1 | - | 5, 6 | 1195 | |||
BC039955.1 | - | 5, 6 | 1138 | 4236297 |