Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002690.2 | NP_002681.1 | 4 | 411 | ||
XM_005273535.3 | XP_005273592.1 | 4 | 408 | |||
XM_005273536.3 | XP_005273593.1 | 4 | 408 | |||
XM_005273537.3 | XP_005273594.1 | 4 | 408 | |||
XM_005273538.2 | XP_005273595.1 | 3 | 318 | |||
XM_005273539.2 | XP_005273596.1 | 3 | 318 | |||
GenBank mRNA | AK223537.1 | - | 4 | 410 | ||
AK314976.1 | - | 4 | 372 | |||
BC100288.1 | - | 4 | 375 | 6151069 | ||
BC106909.1 | - | 4 | 337 | 40031807 | ||
BP370570.1 | - | 4 | 411 | |||
CR536503.1 | - | 4 | 241 | |||
CR541802.1 | - | 4 | 241 | |||
D29013.1 | - | 4 | 354 | |||
JQ664683.1 | - | 4 | 241 | |||
L11607.1 | - | 4 | 241 | |||
M13140.1 | - | 4 | 354 |