Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dkk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145592.2 | NP_663567.1 | 2 | 292 | ||
GenBank mRNA | BC018400.1 | - | 2 | 292 | 3675956 |