Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tti2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199988.1 | NP_001186917.1 | 5 | 1771 | ||
NM_144927.3 | NP_659176.2 | 5 | 1771 | |||
GenBank mRNA | AK045392.1 | - | 5 | 1776 | ||
AK046031.1 | - | 5 | 1761 | |||
AK084242.1 | - | 5 | 1761 | |||
AK139425.1 | - | 5 | 1748 | |||
BC019943.1 | - | 5 | 1474 | 4017819 |