Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127218.2 | NP_001120690.1 | 8, 9 | 1301 | ||
NM_001256879.1 | NP_001243808.1 | 9, 10 | 1662 | |||
NM_006230.3 | NP_006221.2 | 8, 9 | 1128 | |||
XM_011515431.1 | XP_011513733.1 | 8, 9 | 1124 | |||
GenBank mRNA | AK292347.1 | - | 9, 10 | 1662 | ||
AK298168.1 | - | 8, 9 | 992 | |||
AK312294.1 | - | 8, 9 | 1261 | |||
BC000459.2 | - | 8, 9 | 1091 | 2822169 | ||
BC030001.2 | - | 7, 8 | 1068 | |||
U21090.1 | - | 8, 9 | 1090 |