Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csgalnact1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252623.1 | NP_001239552.1 | 9 | 2150 | ||
NM_172753.5 | NP_766341.4 | 8 | 1846 | |||
GenBank mRNA | AK028688.1 | - | 8 | 1769 | ||
AK033522.1 | - | 6 | 1500 | |||
AK080687.1 | - | 8 | 1815 | |||
AK133567.1 | - | 6 | 1499 | |||
AK144089.1 | - | 5 | 1407 | |||
BC026599.1 | - | 3 | 266 | |||
BC057630.1 | - | 10 | 2118 | |||
BC089162.1 | - | 8 | 1843 | 30470205 |