Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006231.3 | NP_006222.2 | 33 | 4386 | ||
XM_011534795.2 | XP_011533097.1 | 33 | 4381 | |||
XM_011534797.2 | XP_011533099.1 | 25 | 3350 | |||
XM_011534798.1 | XP_011533100.1 | 21 | 2902 | |||
XM_011534799.2 | XP_011533101.1 | 33 | 4381 | |||
XM_011534800.2 | XP_011533102.1 | 33 | 4381 | |||
XM_011534802.2 | XP_011533104.1 | 9 | 1203 | |||
GenBank mRNA | BC144561.1 | - | 33 | 4893 | ||
BX647647.1 | - | 7 | 1831 | |||
L09561.1 | - | 33 | 4221 | |||
S60080.1 | - | 32 | 4103 | |||
U49356.1 | - | 33 | 4174 |