Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006231.3 | NP_006222.2 | 2, 3 | 420 | ||
XM_011534795.2 | XP_011533097.1 | 2, 3 | 415 | |||
XM_011534799.2 | XP_011533101.1 | 2, 3 | 415 | |||
XM_011534800.2 | XP_011533102.1 | 2, 3 | 415 | |||
GenBank mRNA | BC144561.1 | - | 2, 3 | 243 | ||
L09561.1 | - | 2, 3 | 255 | |||
U49356.1 | - | 2, 3 | 211 |