Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024922.2 | NP_001020093.2 | 2 | 211 | ||
GenBank mRNA | AK139073.1 | - | 2 | 223 | ||
BC085301.1 | - | 2 | 211 | |||
BC099554.1 | - | 2 | 218 | 30308124 |