Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nxnl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145598.2 | NP_663573.1 | 1 | 113 | ||
GenBank mRNA | BC017153.1 | - | 1 | 87 | ||
BC021911.1 | - | 1 | 65 | 4502967 | ||
BI738445.1 | - | 1 | 113 |