Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001197330.1 | NP_001184259.1 | 2, 3 | 348 | ||
NM_001197331.1 | NP_001184260.1 | 2, 3 | 348 | |||
NM_002692.3 | NP_002683.2 | 2, 3 | 348 | |||
XM_011536842.2 | XP_011535144.1 | 2, 3 | 385 | |||
XM_011536843.2 | XP_011535145.1 | 2, 3 | 181 | |||
XM_011536844.1 | XP_011535146.1 | 2, 3 | 182 | |||
XM_017021374.1 | XP_016876863.1 | 2, 3 | 182 | |||
XM_017021375.1 | XP_016876864.1 | 2, 3 | 181 | |||
GenBank mRNA | AF025840.1 | - | 2, 3 | 301 | ||
AF036899.1 | - | 2, 3 | 185 | |||
AK293163.1 | - | 2, 3 | 180 | |||
AK314963.1 | - | 2, 3 | 183 | |||
BC112962.1 | - | 2, 3 | 178 | 7481939 | ||
BC126218.1 | - | 2, 3 | 271 | 8991934 | ||
BC126220.1 | - | 2, 3 | 271 | 8991936 | ||
BX248064.1 | - | 2, 3 | 186 |