Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rtbdn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144929.2 | NP_659178.1 | 3 | 983 | ||
GenBank mRNA | AK039726.1 | - | 1 | 1490 | ||
AK080715.1 | - | 3 | 299 | |||
AY428631.1 | - | 2 | 195 | |||
AY527275.1 | - | 3 | 983 | |||
AY527276.1 | - | 4 | 438 | |||
BC022744.1 | - | 2 | 555 | 4527012 | ||
BC023051.1 | - | 2 | 511 | 5361063 | ||
BC027079.1 | - | 2 | 511 | 5357069 |