Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Heatr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172757.3 | NP_766345.3 | 10 | 1465 | ||
GenBank mRNA | AK047782.1 | - | 10 | 1496 | ||
AK052635.1 | - | 10 | 1492 | |||
AK134510.1 | - | 10 | 1470 | |||
AK136424.1 | - | 10 | 1475 | |||
AK140522.1 | - | 10 | 1483 | |||
AK163715.1 | - | 10 | 2228 | |||
AK165541.1 | - | 10 | 1359 | |||
BC030373.1 | - | 6 | 810 | |||
BC065060.1 | - | 10 | 1465 | 6402217 |