Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc38a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172758.4 | NP_766346.1 | 7, 8 | 1023 | ||
GenBank mRNA | AK052548.1 | - | 7, 8 | 1024 | ||
AK171137.1 | - | 7, 8 | 1031 | |||
AK194183.1 | - | 2, 3 | 139 | |||
BC031853.1 | - | 5, 6 | 898 | 4009949 |