Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dync1li2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013380.2 | NP_001013398.2 | 7 | 1112 | ||
GenBank mRNA | AK140743.1 | - | 7 | 880 | ||
AK147899.1 | - | 7 | 879 | |||
AK152157.1 | - | 7 | 877 | |||
AK161434.1 | - | 7 | 974 | |||
AK161494.1 | - | 7 | 974 | |||
BC058645.1 | - | 7 | 1070 | 6405596 |