Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ces2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145603.2 | NP_663578.1 | 10 | 1397 | ||
GenBank mRNA | AB110073.1 | - | 10 | 1397 | ||
AK136411.1 | - | 10 | 1397 | |||
BC015290.1 | - | 10 | 1381 | 4241028 | ||
BC024552.1 | - | 10 | 1379 | 4990820 | ||
BC031170.1 | - | 10 | 1379 | 4984806 | ||
BC034178.1 | - | 10 | 1366 | 4971285 | ||
BC034180.1 | - | 10 | 1379 | 4972755 | ||
BC034182.1 | - | 6 | 697 | |||
BC034191.1 | - | 10 | 1379 | 4982995 |
Gene Symbol: Ces2d-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AA734872.1 | - | 180 | |||
BF721059.1 | - | 93 |