Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ces2e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163756.1 | NP_001157228.1 | 10 | 1470 | ||
NM_172759.3 | NP_766347.1 | 10 | 1470 | |||
GenBank mRNA | AK033563.1 | - | 10 | 1433 | ||
AK077248.1 | - | 10 | 1461 | |||
BC022148.1 | - | 6 | 783 | |||
BC055062.1 | - | 10 | 1439 | 6528966 |