Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Edc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301100.1 | NP_001288029.1 | 9 | 1257 | ||
NM_181594.3 | NP_853625.1 | 9 | 1257 | |||
GenBank mRNA | AK146527.1 | - | 9 | 1223 | ||
AK152963.1 | - | 9 | 1234 | |||
BC053081.1 | - | 9 | 1187 | 5704584 |