Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vac14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146216.2 | NP_666328.2 | 14 | 1819 | ||
GenBank mRNA | AK031952.1 | - | 14 | 1854 | ||
AK043319.1 | - | 14 | 1851 | |||
AK046826.1 | - | 14 | 1791 | |||
AK054468.1 | - | 5 | 646 | |||
AK084520.1 | - | 14 | 1870 | |||
AK140191.1 | - | 13 | 1689 | |||
AK171577.1 | - | 14 | 1859 | |||
AK171883.1 | - | 14 | 1847 | |||
AK191163.1 | - | 3 | 256 | |||
AK205121.1 | - | 2 | 68 | |||
AK211785.1 | - | 2 | 68 | |||
AK217344.1 | - | 2 | 68 | |||
BC032215.1 | - | 7 | 732 | |||
BC052199.1 | - | 14 | 1819 | 6512678 |