Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fuk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172283.3 | NP_758487.2 | 11 | 1075 | ||
GenBank mRNA | AJ297482.4 | - | 11 | 1046 | ||
AJ534942.1 | - | 11 | 1046 | |||
AK131165.1 | - | 3 | 755 | |||
AK161763.1 | - | 11 | 1077 | |||
AK164340.1 | - | 11 | 1044 | |||
AK183286.1 | - | 1 | 14 | |||
AK197389.1 | - | 3 | 280 | |||
AY223865.1 | - | 11 | 1046 | |||
BC037698.1 | - | 10 | 1024 | 4190449 |