Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fuk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172283.3 | NP_758487.2 | 19 | 2547 | ||
GenBank mRNA | AJ297482.4 | - | 19 | 2518 | ||
AJ534942.1 | - | 19 | 2518 | |||
AK049153.1 | - | 8 | 1376 | |||
AK131165.1 | - | 8 | 3897 | |||
AK161763.1 | - | 19 | 2651 | |||
AK164340.1 | - | 19 | 2614 | |||
AY223865.1 | - | 19 | 2518 | |||
BC037698.1 | - | 18 | 2496 | 4190449 |