Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem231 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033321.1 | NP_001028493.1 | 5 | 621 | ||
GenBank mRNA | AK030046.1 | - | 6 | 710 | ||
AK155425.1 | - | 5 | 621 | |||
BC100402.1 | - | 5 | 605 | 30106220 | ||
BC132428.1 | - | 5 | 711 | 40130705 | ||
BC132430.1 | - | 5 | 711 | 40130707 |