Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Piezo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037298.1 | NP_001032375.1 | 24, 25 | 3736 | ||
GenBank mRNA | AK129095.1 | - | 2, 3 | 388 | ||
BC039210.1 | - | 3, 4 | 485 | |||
BC042207.1 | - | 3, 4 | 485 | |||
BC044757.2 | - | 10, 11 | 1498 | |||
HQ215520.1 | - | 24, 25 | 3448 |