Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Piezo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037298.1 | NP_001032375.1 | 14 | 2042 | ||
GenBank mRNA | HQ215520.1 | - | 14 | 1754 |