Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLR2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000938.2 | NP_000929.1 | 24 | 3699 | ||
NM_001303268.1 | NP_001290197.1 | 23 | 3548 | |||
NM_001303269.1 | NP_001290198.1 | 25 | 3877 | |||
GenBank mRNA | AF055028.1 | - | 10 | 1259 | ||
AK098271.1 | - | 1 | 1035 | |||
AK289823.1 | - | 24 | 3443 | |||
AK294901.1 | - | 23 | 3324 | |||
AK295132.1 | - | 25 | 3648 | |||
AK299090.1 | - | 24 | 3215 | |||
BC023503.2 | - | 24 | 3457 | 3346389 | ||
X63563.1 | - | 24 | 3396 |