Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145607.3 | NP_663582.2 | 16 | 1835 | ||
GenBank mRNA | AK036992.1 | - | 2 | 3754 | ||
AK144842.1 | - | 16 | 1836 | |||
AK148102.1 | - | 18 | 2065 | |||
AK160894.1 | - | 18 | 2072 | |||
BC017545.1 | - | 10 | 1046 | |||
BC072577.1 | - | 16 | 1825 | |||
BC079851.1 | - | 17 | 1890 | 6401930 |