Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B130055M24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK156871.1 | - | 224 |
Gene Symbol: Ppan | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145610.2 | NP_663585.2 | 2, 3 | 315 | ||
GenBank mRNA | AK039027.1 | - | 2, 3 | 220 | ||
AK171672.1 | - | 1, 2 | 309 | |||
AK172615.1 | - | 1, 2 | 309 | |||
BC014688.1 | - | 2, 3 | 233 | 4489442 | ||
BC059277.1 | - | 2, 3 | 205 | |||
CJ077201.1 | - | 2, 3 | 316 |