Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scn3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083917.1 | NP_001077386.1 | 3, 4 | 629 | ||
NM_001286614.1 | NP_001273543.1 | 3, 4 | 625 | |||
NM_178227.4 | NP_839941.1 | 3, 4 | 629 | |||
GenBank mRNA | AK049286.1 | - | 3, 4 | 627 | ||
AK049747.1 | - | 3, 4 | 448 | |||
AK076466.1 | - | 2, 3 | 273 | |||
AK173115.1 | - | 3, 4 | 506 | |||
AY049036.1 | - | 2, 3 | 234 | |||
BC032253.1 | - | 3, 4 | 423 | 5368626 | ||
BC053919.1 | - | 2, 3 | 524 | 6308278 | ||
BC058083.1 | - | 3, 4 | 568 | |||
BC058636.1 | - | 3, 4 | 628 | 5702879 | ||
CB520894.1 | - | 3, 4 | 629 |