Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem136 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034863.3 | NP_001030035.1 | 3 | 750 | ||
GenBank mRNA | AK128910.1 | - | 3 | 799 | ||
AK158698.1 | - | 3 | 751 | |||
AK214821.1 | - | 1 | 217 | |||
BC139046.1 | - | 3 | 737 | 8862068 | ||
BC139047.1 | - | 3 | 737 | 8862069 |