Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175482.3 | NP_780691.2 | 16 | 1914 | ||
GenBank mRNA | AK028335.1 | - | 2 | 109 | ||
AK036673.1 | - | 6 | 687 | |||
AK052442.1 | - | 8 | 947 | |||
AK129380.1 | - | 6 | 647 | |||
BC066033.1 | - | 16 | 1810 | 6835629 | ||
BC088733.1 | - | 16 | 1836 | 30459522 |