Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175482.3 | NP_780691.2 | 21 | 2697 | ||
GenBank mRNA | AK028335.1 | - | 7 | 892 | ||
AK052442.1 | - | 13 | 1730 | |||
AK129380.1 | - | 11 | 1430 | |||
BC066033.1 | - | 21 | 2593 | 6835629 | ||
BC088733.1 | - | 21 | 2619 | 30459522 | ||
BG087938.2 | - | 5 | 527 |