Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam214a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113283.1 | NP_001106754.1 | 9 | 2887 | ||
NM_153584.2 | NP_705812.2 | 8 | 2710 | |||
GenBank mRNA | AK173161.1 | - | 9 | 2758 | ||
BC031353.1 | - | 8 | 2710 | 5363230 | ||
BC032966.1 | - | 4 | 481 | |||
BC060215.1 | - | 9 | 2885 |