Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd109 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153098.3 | NP_694738.1 | 11 | 1587 | ||
GenBank mRNA | AK029247.1 | - | 11 | 1361 | ||
AK036799.1 | - | 11 | 1351 | |||
AY083458.1 | - | 11 | 1593 | |||
BC052443.1 | - | 11 | 1355 | 6406841 | ||
DQ832284.1 | - | 11 | 1593 |