Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camkv | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145621.2 | NP_663596.1 | 4 | 425 | ||
GenBank mRNA | AK085143.1 | - | 431 | |||
AK147323.1 | - | 4 | 413 | |||
BC017634.1 | - | 4 | 425 | 4527857 | ||
BC022169.1 | - | 4 | 420 | |||
BC086762.1 | - | 4 | 409 | |||
BC111033.1 | - | 4 | 420 | 5368225 |