Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Als2cl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146059.1 | NP_001139531.1 | 17 | 1882 | ||
NM_001146060.1 | NP_001139532.1 | 17 | 1962 | |||
NM_146228.4 | NP_666340.3 | 17 | 1879 | |||
GenBank mRNA | AB107016.1 | - | 17 | 1879 | ||
AF481142.1 | - | 16 | 1775 | |||
AK030815.1 | - | 10 | 1016 | |||
AK053076.1 | - | 17 | 1852 | |||
AK131149.1 | - | 17 | 1881 | |||
AK155451.1 | - | 17 | 1948 | |||
BC117928.1 | - | 16 | 1832 | 40097635 | ||
BC117929.1 | - | 16 | 1829 | 40097636 |