Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rtp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153100.2 | NP_694740.2 | 3 | 992 | ||
GenBank mRNA | AJ428064.1 | - | 3 | 992 | ||
AY562227.1 | - | 3 | 890 | |||
BC112903.1 | - | 3 | 992 | 4196964 | ||
BC150678.1 | - | 3 | 936 | 9087589 |