Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pyhin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175026.3 | NP_778191.1 | 6 | 1490 | ||
GenBank mRNA | AK076726.1 | - | 2 | 368 | ||
AK080371.1 | - | 6 | 1480 | |||
AK150817.1 | - | 6 | 1381 | |||
BC096384.1 | - | 6 | 1444 | 30752094 | ||
BC107378.1 | - | 7 | 1432 | 40056703 | ||
BC107379.1 | - | 6 | 1324 | 40056704 |