Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PON2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000305.2 | NP_000296.2 | 6 | 731 | ||
NM_001018161.1 | NP_001018171.1 | 6 | 695 | |||
XM_005250453.1 | XP_005250510.1 | 6 | 661 | |||
XM_017012357.1 | XP_016867846.1 | 5 | 580 | |||
XM_017012358.1 | XP_016867847.1 | 5 | 606 | |||
GenBank mRNA | AB102891.1 | - | 6 | 574 | ||
AF001601.1 | - | 6 | 642 | |||
AF001602.1 | - | 6 | 653 | |||
AK291103.1 | - | 6 | 653 | |||
AK296029.1 | - | 6 | 610 | |||
AK296636.1 | - | 6 | 651 | |||
AK301313.1 | - | 6 | 858 | |||
AK308065.1 | - | 6 | 662 | |||
AK315209.1 | - | 6 | 679 | |||
BC040010.1 | - | 6 | 634 | 6056277 | ||
BC046160.1 | - | 6 | 650 | 5393700 | ||
L48513.1 | - | 6 | 627 |