Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167775.1 | NP_001161247.1 | 5 | 1159 | ||
NM_001167776.1 | NP_001161248.1 | 4 | 641 | |||
NM_145627.2 | NP_663602.1 | 5 | 872 | |||
GenBank mRNA | AK041871.1 | - | 4 | 616 | ||
AK089105.1 | - | 4 | 452 | |||
AK122208.1 | - | 4 | 559 | |||
AK145991.1 | - | 5 | 872 | |||
AK153736.1 | - | 5 | 1160 | |||
AK167666.1 | - | 4 | 619 | |||
BC004674.1 | - | 4 | 557 | 3500403 | ||
BC093519.1 | - | 4 | 573 |