Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmgt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146234.3 | NP_666346.1 | 2 | 173 | ||
GenBank mRNA | AK036253.1 | - | 2 | 174 | ||
AK043524.1 | - | 2 | 187 | |||
AK049689.1 | - | 2 | 148 | |||
AK163269.1 | - | 2 | 325 | |||
BC032271.1 | - | 2 | 199 | 4954726 | ||
EU069461.1 | - | 2 | 132 |