Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BC023829 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033328.2 | NP_001028500.1 | 5, 6 | 762 | ||
GenBank mRNA | AK158294.1 | - | 5, 6 | 762 | ||
BC023829.1 | - | 2, 3 | 217 | |||
BC138670.1 | - | 6, 7 | 842 | 8861691 | ||
BC145368.1 | - | 6, 7 | 842 | 9053926 |