Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrp11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172784.3 | NP_766372.1 | 3 | 1015 | ||
GenBank mRNA | AK031994.1 | - | 3 | 957 | ||
AK036676.1 | - | 3 | 998 | |||
AK048919.1 | - | 3 | 1031 | |||
AK050544.1 | - | 3 | 985 | |||
BC059874.1 | - | 3 | 836 |