Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il20ra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172786.2 | NP_766374.1 | 7 | 968 | ||
GenBank mRNA | AK054215.1 | - | 7 | 969 | ||
BC056628.1 | - | 7 | 952 | 5033177 |