Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: L3mbtl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172787.2 | NP_766375.1 | 6 | 595 | ||
GenBank mRNA | AK031398.1 | - | 6 | 581 | ||
AK045667.1 | - | 6 | 596 | |||
AK049597.1 | - | 6 | 630 | |||
AK129443.1 | - | 2 | 140 | |||
BC082309.1 | - | 5 | 465 | 30356672 | ||
BC085192.1 | - | 5 | 700 | 30694487 |